Anti-SMOX Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35213-50UG A35213-100UG
77800-354EA 355.5 CAD
77800-354 77800-353
Anti-SMOX Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SMOX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR