Anti-TBCC Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A36209-100UG A36209-50UG
77801-937EA 521.4 CAD
77801-937 77801-938
Anti-TBCC Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TBCC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR