Anti-PKD1L2 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A59754-100UG A59754-50UG
77832-490EA 521.4 CAD
77832-490 77832-491
Anti-PKD1L2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PKD1L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR