Anti-MMP2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A42126-100UG A42126-50UG
77810-152EA 521.4 CAD
77810-152 77810-153
Anti-MMP2 Rabbit Recombinant Polyclonal Antibody
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: MMP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR