Anti-PIGZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A59699-100UG A59699-50UG
77832-386EA 521.4 CAD
77832-386 77832-387
Anti-PIGZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PIGZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR