Anti-NUDT9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A69297-100UG A69297-50UG
77840-336EA 521.4 CAD
77840-336 77840-337
Anti-NUDT9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: NUDT9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR