Anti-GATD3A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31388-100UG A31388-50UG
77794-411EA 521.4 CAD
77794-411 77794-412
Anti-GATD3A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: GATD3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR