Anti-GK3P Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A54441-100UG A54441-50UG
77822-500EA 521.4 CAD
77822-500 77822-501
Anti-GK3P Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GK3P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR