Anti-GCY1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50425-100UG A50425-50UG
77815-266EA 521.4 CAD
77815-266 77815-267
Anti-GCY1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GCY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR