Anti-PCF11 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A30752-100UG A30752-50UG
77793-394EA 521.4 CAD
77793-394 77793-395
Anti-PCF11 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: PCF11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PCF11
Environmentally Preferable
Antigen name Pre-mRNA cleavage complex 2 protein Pcf11 protein (409-604AA)
Antigen synonyms hPCF11|KIAA0824|OTTHUMP00000234826|PCF11|PCF11 cleavage and polyadenylation factor subunit|cleavage and polyadenylation factor II subunit|homolog|cleavage and polyadenylation factor subunit|homolog (S.|PCF11_HUMAN|PCF11p homolog|Pre-mRNA cleavage complex 2 protein Pcf11|Pre-mRNA cleavage complex II protein Pcf11
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID O94913
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR