Anti-SMUG1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A57185-50UG A57185-100UG
77827-652EA 355.5 CAD
77827-652 77827-651
Anti-SMUG1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: SMUG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR