Anti-Major pollen allergen Dac g 4 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A50132-100UG A50132-50UG
77814-709EA 521.4 CAD
77814-709 77814-710
Anti-Major pollen allergen Dac g 4 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Major pollen allergen Dac g 4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dactylis glomeRata
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR