Anti-POMGNT1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65784-100UG A65784-50UG
77833-732EA 521.4 CAD
77833-732 77833-733
Anti-POMGNT1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: POMGNT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR