Anti-PPP1R3B Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A59510-100UG A59510-50UG
77832-028EA 521.4 CAD
77832-028 77832-029
Anti-PPP1R3B Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PPP1R3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR