Anti-SLC12A5 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A67060-100UG A67060-50UG
77836-134EA 521.4 CAD
77836-134 77836-135
Anti-SLC12A5 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:500 to 1:1000, IF:1:50 to 1:200

Type:
Antigen: SLC12A5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR