Anti-PIGV Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A67769-100UG A67769-50UG
77837-471EA 521.4 CAD
77837-471 77837-472
Anti-PIGV Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PIGV
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR