Anti-POLG Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A31573-100UG A31573-50UG
77794-705EA 521.4 CAD
77794-705 77794-706
Anti-POLG Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: POLG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR