Anti-GK5 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A58345-100UG A58345-50UG
77829-833EA 521.4 CAD
77829-833 77829-834
Anti-GK5 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GK5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR