Anti-PDRG1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A69051-100UG A69051-50UG
77839-869EA 521.4 CAD
77839-869 77839-870
Anti-PDRG1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: PDRG1
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR