Anti-MTR Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66428-100UG A66428-50UG
77834-945EA 521.4 CAD
77834-945 77834-946
Anti-MTR Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MTR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR