Anti-OPN3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A68211-50UG A68211-100UG
77838-298EA 355.5 CAD
77838-298 77838-297
Anti-OPN3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: OPN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR