Anti-HECTD3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A57668-100UG A57668-50UG
77828-555EA 521.4 CAD
77828-555 77828-556
Anti-HECTD3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: HECTD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR