Anti-cpg2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51614-100UG A51614-50UG
77817-504EA 521.4 CAD
77817-504 77817-505
Anti-cpg2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: cpg2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas sp.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR