Anti-TES-26 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51139-100UG A51139-50UG
77816-611EA 521.4 CAD
77816-611 77816-612
Anti-TES-26 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TES-26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Toxocara canis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR