Anti-PDGFRA Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A30861-100UG A30861-50UG
77793-573EA 521.4 CAD
77793-573 77793-574
Anti-PDGFRA Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: PDGFRA
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol PDGFRA
Antigen name Platelet-derived growth factor receptor alpha protein (30-198AA)
Environmentally Preferable
Antigen synonyms Alpha-type platelet-derived growth factor receptor|CD140 antigen-like family member A|CD140a|CD140a antigen|MGC74795|PDGF alpha chain|PDGF Receptor alpha|PDGF-R-alpha|PDGFR 2|PDGFR alpha|PDGFR2|PDGFRA|PDGFRA/BCR fusion|PGFRA_HUMAN|Platelet derived growth factor receptor 2|Platelet derived growth factor receptor alpha|Platelet derived growth factor receptor alpha polypeptide|Platelet derived growth factor receptor|Rearranged in hypereosinophilia platelet derived growth factor receptor alpha fusion protein|RHEPDGFRA
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P16234
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR