Anti-NAT10 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A67019-100UG A67019-50UG
77836-056EA 521.4 CAD
77836-056 77836-057
Anti-NAT10 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: NAT10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR