Anti-Major pepsin inhibitor 3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50483-100UG A50483-50UG
77815-377EA 521.4 CAD
77815-377 77815-378
Anti-Major pepsin inhibitor 3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Major pepsin inhibitor 3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Ascaris suum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR