Anti-PDE1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A49455-100UL A49455-50UL
77813-540EA 521.4 CAD
77813-540 77813-541
Anti-PDE1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PDE1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR