Anti-glpD Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50383-100UG A50383-50UG
77815-185EA 521.4 CAD
77815-185 77815-186
Anti-glpD Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: glpD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR