Anti-FAM153C Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A56591-100UG A56591-50UG
77826-566EA 521.4 CAD
77826-566 77826-567
Anti-FAM153C Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: FAM153C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR