Anti-SMPD2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35224-50UG A35224-100UG
77800-374EA 355.5 CAD
77800-374 77800-373
Anti-SMPD2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SMPD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR