Anti-MIOS Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A67309-100UL A67309-50UL
77836-604EA 521.4 CAD
77836-604 77836-605
Anti-MIOS Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MIOS
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR