Anti-OXSM Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A67809-100UG A67809-50UG
77837-548EA 521.4 CAD
77837-548 77837-549
Anti-OXSM Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OXSM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR