Anti-DEFA1/DEFA3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52263-100UL A52263-50UL
77818-650EA 521.4 CAD
77818-650 77818-651
Anti-DEFA1/DEFA3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: DEFA1/DEFA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR