Anti-ABHD10 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A67271-100UG A67271-50UG
77836-533EA 521.4 CAD
77836-533 77836-534
Anti-ABHD10 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ABHD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR