Anti-GCY1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50421-100UG A50421-50UG
77815-258EA 521.4 CAD
77815-258 77815-259
Anti-GCY1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GCY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR