Anti-MRPL22 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A69073-100UG A69073-50UG
77839-910EA 521.4 CAD
77839-910 77839-911
Anti-MRPL22 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MRPL22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR