Anti-RHBDL2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A33193-100UG A33193-50UG
77797-261EA 521.4 CAD
77797-261 77797-262
Anti-RHBDL2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: RHBDL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR