Anti-TERT Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A36385-100UG A36385-50UG
77802-226EA 521.4 CAD
77802-226 77802-227
Anti-TERT Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: TERT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR