Anti-AES Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52948-100UL A52948-50UL
77819-896EA 521.4 CAD
77819-896 77819-897
Anti-AES Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: AES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR