Anti-ERO1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A59908-100UG A59908-50UG
77832-777EA 521.4 CAD
77832-777 77832-778
Anti-ERO1B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ERO1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR