Anti-UTS2R Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49323-50UL A49323-100UL
77813-326EA 355.5 CAD
77813-326 77813-325
Anti-UTS2R Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: UTS2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR