Anti-Histidine-rich protein PFHRP-II Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51337-100UG A51337-50UG
77816-988EA 521.4 CAD
77816-988 77816-989
Anti-Histidine-rich protein PFHRP-II Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Histidine-rich protein PFHRP-II
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR