Anti-BCAT1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52139-100UL A52139-50UL
77818-457EA 521.4 CAD
77818-457 77818-458
Anti-BCAT1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: BCAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR