Anti-ZNF454 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A38976-50UG A38976-100UG
77805-480EA 355.5 CAD
77805-480 77805-479
Anti-ZNF454 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ZNF454
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR