Anti-USP28 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49285-100UL A49285-50UL
77813-262EA 521.4 CAD
77813-262 77813-263
Anti-USP28 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: USP28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR