Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A69497-50UG A69497-100UG
77840-718EA 355.5 CAD
77840-718 77840-717
Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: DCLRE1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR