Anti-AIFM2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A68620-100UL A68620-50UL
77839-067EA 521.4 CAD
77839-067 77839-068
Anti-AIFM2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: AIFM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR