Anti-egsA Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A52346-100UG A52346-50UG
77818-796EA 521.4 CAD
77818-796 77818-797
Anti-egsA Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: egsA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Geobacillus stearothermophilus
New Product
Order Now

SPECIFICATIONS

Antigen symbol egsA
Environmentally Preferable
Antigen name Recombinant Geobacillus stearothermophilus Glycerol-1-phosphate dehydrogenase [NAD(P)+] protein (1-404AA)
Antigen synonyms egsA|araMGlycerol-1-phosphate dehydrogenase [NAD(P)+]|G1P dehydrogenase|G1PDH|EC 1.1.1.261|Enantiomeric glycerophosphate synthase|sn-glycerol-1-phosphate dehydrogenase
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Geobacillus stearothermophilus
Host Rabbit
Gene ID B3EYN8
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR