Anti-botB Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A50295-100UG A50295-50UG
77815-014EA 521.4 CAD
77815-014 77815-015
Anti-botB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: botB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR